Assay Details
| Gene Symbol: RGS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454858.1 | XP_015310344.1 | 3 - 4 | 1249 | 151 | |
| XM_015454859.1 | XP_015310345.1 | 4 - 5 | 1942 | 151 | ||
| GenBank mRNA | AB220555.1 | - | 2 - 3 | 464 | 151 | |
More Information
Set Membership: |
|

RGS20
Genomic Map