Assay Details
| Gene Symbol: CTSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582086.2 | XP_005582143.1 | 7 - 8 | 1787 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTSL| Gene Symbol: CTSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582086.2 | XP_005582143.1 | 7 - 8 | 1787 | 87 | |
Set Membership: |
|