Assay Details
| Gene Symbol: CYTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573223.2 | XP_005573280.1 | 6 - 7 | 627 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYTIP| Gene Symbol: CYTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573223.2 | XP_005573280.1 | 6 - 7 | 627 | 81 | |
Set Membership: |
|