Assay Details
| Gene Symbol: LOC102147203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542801.2 | XP_005542858.1 | 1 - 2 | 970 | 73 | |
| GenBank mRNA | AB170879.1 | - | 1 - 2 | 48 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005542801.2 transcripts.
Set Membership: |
|

LOC102147203
Genomic Map