Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549866.2 | XP_005549923.1 | 7 - 8 | 1356 | 74 | |
| XM_005549867.2 | XP_005549924.1 | 7 - 8 | 1356 | 74 | ||
| XM_005549868.2 | XP_005549925.1 | 7 - 8 | 1356 | 74 | ||
| XM_015447574.1 | XP_015303060.1 | 7 - 8 | 1356 | 74 | ||
| XM_015447575.1 | XP_015303061.1 | 7 - 8 | 1356 | 74 | ||
| XM_015447576.1 | XP_015303062.1 | 7 - 8 | 1356 | 74 | ||
| XM_015447577.1 | XP_015303063.1 | 7 - 8 | 1356 | 74 | ||
| XM_015447578.1 | XP_015303064.1 | 7 - 8 | 1356 | 74 | ||
More Information
Set Membership: |
|

MINDY4
Genomic Map