Assay Details
| Gene Symbol: POP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588689.2 | XP_005588746.1 | 5 - 6 | 490 | 140 | |
| XM_015441842.1 | XP_015297328.1 | 5 - 6 | 485 | 140 | ||
More Information
Set Membership: |
|

POP4
Genomic Map