Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015450060.1 | XP_015305546.1 | 1 - 2 | 351 | 110 | |
| GenBank mRNA | AB170138.1 | - | 1 - 2 | 219 | 110 | |
More Information
Set Membership: |
|

LRPAP1
Genomic Map