Assay Details
| Gene Symbol: METTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545721.2 | XP_005545778.1 | 2 - 3 | 473 | 76 | |
| XM_015445062.1 | XP_015300548.1 | 2 - 3 | 464 | 76 | ||
| XM_015445063.1 | XP_015300549.1 | 2 - 3 | 420 | 76 | ||
| XM_015445064.1 | XP_015300550.1 | 2 - 3 | 473 | 76 | ||
| XM_015445066.1 | XP_015300552.1 | 2 - 3 | 473 | 76 | ||
| XM_015445067.1 | XP_015300553.1 | 2 - 3 | 473 | 76 | ||
| XM_015445068.1 | XP_015300554.1 | 2 - 3 | 473 | 76 | ||
| XM_015445069.1 | XP_015300555.1 | 2 - 3 | 483 | 76 | ||
More Information
Set Membership: |
|

METTL6
Genomic Map