Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561313.2 | XP_005561370.1 | 6 - 7 | 811 | 92 | |
| XM_005561314.1 | XP_005561371.1 | 5 - 6 | 674 | 92 | ||
More Information
Set Membership: |
|

LGALS3
Genomic Map