Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555943.2 | XP_005556000.1 | 1 - 2 | 622 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555943.2 | XP_005556000.1 | 1 - 2 | 622 | 122 |
Set Membership: |
![]() ![]() ![]() ![]() |