Assay Details
| Gene Symbol: SOHLH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585636.2 | XP_005585693.1 | 1 - 2 | 362 | 69 | |
| XM_015439148.1 | XP_015294634.1 | 1 - 2 | 365 | 69 | ||
More Information
Set Membership: |
|

SOHLH2
Genomic Map