Assay Details
| Gene Symbol: SH3GL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581667.1 | XP_005581724.1 | 8 - 9 | 874 | 120 | |
| XM_015437118.1 | XP_015292604.1 | 8 - 9 | 915 | 120 | ||
| XM_015437119.1 | XP_015292605.1 | 8 - 9 | 980 | 120 | ||
More Information
Set Membership: |
|

SH3GL2
Genomic Map