Assay Details
| Gene Symbol: MBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586396.2 | XP_005586453.1 | 9 - 10 | 1177 | 71 | |
| XM_005586397.2 | XP_005586454.1 | 8 - 9 | 1146 | 71 | ||
| XM_005586399.2 | XP_005586456.1 | 7 - 8 | 1065 | 71 | ||
| XM_015439600.1 | XP_015295086.1 | 8 - 9 | 1096 | 71 | ||
| GenBank mRNA | AB170099.1 | - | 4 - 5 | 486 | 71 | |
| AB170192.1 | - | 3 - 4 | 290 | 71 | ||
More Information
Set Membership: |
|

MBP
Genomic Map