Assay Details
| Gene Symbol: DKK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285227.1 | NP_001272156.1 | 4 - 5 | 501 | 57 | |
| XM_015434119.1 | XP_015289605.1 | 4 - 5 | 653 | 57 | ||
| XM_015434120.1 | XP_015289606.1 | 4 - 5 | 706 | 57 | ||
| GenBank mRNA | AB169907.1 | - | 4 - 5 | 501 | 57 | |
More Information
Set Membership: |
|

DKK3
Genomic Map