Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588933.2 | XP_005588990.1 | 1 - 2 | 625 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP8| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588933.2 | XP_005588990.1 | 1 - 2 | 625 | 83 | |
Set Membership: |
|