Assay Details
| Gene Symbol: BLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560522.2 | XP_005560579.1 | 2 - 3 | 222 | 123 | |
| XM_015453405.1 | XP_015308891.1 | 3 - 4 | 325 | 123 | ||
More Information
Set Membership: |
|

BLM
Genomic Map