Assay Details
| Gene Symbol: ACADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542992.2 | XP_005543049.1 | 6 - 7 | 922 | 91 | |
| XM_015433969.1 | XP_015289455.1 | 6 - 7 | 716 | 91 | ||
| GenBank mRNA | AB169627.1 | - | 6 - 7 | 497 | 91 | |
| AB170574.1 | - | 6 - 7 | 497 | 91 | ||
| AB171682.1 | - | 6 - 7 | 499 | 91 | ||
More Information
Set Membership: |
|

ACADM
Genomic Map