Assay Details
| Gene Symbol: PPP1R36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561477.2 | XP_005561534.1 | 1 - 2 | 155 | 93 | |
| XM_015453891.1 | XP_015309377.1 | 1 - 2 | 137 | 93 | ||
| GenBank mRNA | AB168605.1 | - | 1 - 2 | 122 | 93 | |
More Information
Set Membership: |
|

PPP1R36
Genomic Map