Assay Details
| Gene Symbol: MEPCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549225.2 | XP_005549282.1 | 2 - 3 | 2675 | 71 | |
| GenBank mRNA | AB179198.1 | - | 3 - 4 | 1436 | 71 | |
More Information
Set Membership: |
|

MEPCE
Genomic Map