Assay Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578641.2 | XP_005578698.1 | 9 - 10 | 1456 | 59 | |
| XM_005578642.2 | XP_005578699.1 | 8 - 9 | 1348 | 59 | ||
More Information
Set Membership: |
|

SWAP70
Genomic Map