Assay Details
| Gene Symbol: LOC102128245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564811.2 | XP_005564868.1 | 4 - 5 | 1049 | 90 | |
| XM_005564812.2 | XP_005564869.1 | 4 - 5 | 1079 | 90 | ||
| XM_015455664.1 | XP_015311150.1 | 3 - 4 | 1809 | 90 | ||
More Information
Set Membership: |
|

LOC102128245
Genomic Map