Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580586.1 | XP_005580643.1 | 1 - 2 | 240 | 63 | |
| XM_005580587.2 | XP_005580644.1 | 1 - 2 | 240 | 63 | ||
| XM_015436306.1 | XP_015291792.1 | 1 - 2 | 240 | 63 | ||
| XM_015436307.1 | XP_015291793.1 | 1 - 2 | 240 | 63 | ||
| XM_015436308.1 | XP_015291794.1 | 1 - 2 | 240 | 63 | ||
More Information
Set Membership: |
|

MED27
Genomic Map