Assay Details
| Gene Symbol: MDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540551.2 | XP_005540608.1 | 6 - 7 | 600 | 91 | |
| XM_005540552.2 | XP_005540609.1 | 6 - 7 | 572 | 91 | ||
| XM_005540553.2 | XP_005540610.1 | 6 - 7 | 601 | 91 | ||
More Information
Set Membership: |
|

MDM4
Genomic Map