Assay Details
| Gene Symbol: RASSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547221.2 | XP_005547278.1 | 4 - 5 | 955 | 79 | |
| XM_005547223.2 | XP_005547280.1 | 3 - 4 | 848 | 79 | ||
| XM_015445941.1 | XP_015301427.1 | 3 - 4 | 1120 | 79 | ||
More Information
Set Membership: |
|

RASSF1
Genomic Map