Assay Details
| Gene Symbol: DES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574383.2 | XP_005574440.2 | 3 - 4 | 918 | 64 | |
| GenBank mRNA | AB168811.1 | - | 3 - 4 | 266 | 64 | |
| AB445461.1 | - | 3 - 4 | 823 | 64 | ||
More Information
Set Membership: |
|

DES
Genomic Map