Assay Details
| Gene Symbol: MAP2K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583146.2 | XP_005583203.1 | 3 - 4 | 396 | 73 | |
| XM_005583148.2 | XP_005583205.1 | 3 - 4 | 421 | 73 | ||
| XM_015437780.1 | XP_015293266.1 | 3 - 4 | 2279 | 73 | ||
| XM_015437782.1 | XP_015293268.1 | 3 - 4 | 138 | 73 | ||
More Information
Set Membership: |
|

MAP2K3
Genomic Map