Assay Details
| Gene Symbol: BPNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540883.2 | XP_005540940.1 | 8 - 9 | 949 | 111 | |
| XM_005540884.2 | XP_005540941.1 | 9 - 10 | 1003 | 111 | ||
| XM_005540885.2 | XP_005540942.1 | 8 - 9 | 948 | 111 | ||
| XM_015450181.1 | XP_015305667.1 | 9 - 10 | 1013 | 111 | ||
| XM_015450197.1 | XP_015305683.1 | 8 - 9 | 985 | 111 | ||
| XM_015450199.1 | XP_015305685.1 | 8 - 9 | 907 | 111 | ||
| XM_015450201.1 | XP_015305687.1 | 7 - 8 | 841 | 111 | ||
| XM_015450205.1 | XP_015305691.1 | 7 - 8 | 878 | 111 | ||
More Information
Set Membership: |
|

BPNT1
Genomic Map