Assay Details
| Gene Symbol: LOC102142548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319501.1 | NP_001306430.1 | 6 - 7 | 632 | 66 | |
| XM_005541934.2 | XP_005541991.1 | 6 - 7 | 774 | 66 | ||
| GenBank mRNA | AB171075.1 | - | 6 - 7 | 632 | 66 | |
More Information
Set Membership: |
|

LOC102142548
Genomic Map