Assay Details
| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580975.2 | XP_005581032.1 | 4 - 5 | 649 | 98 | |
| XM_005580976.2 | XP_005581033.1 | 4 - 5 | 649 | 98 | ||
More Information
Set Membership: |
|

ORM1
Genomic Map