Assay Details
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284635.1 | NP_001271564.1 | 2 - 3 | 228 | 73 | |
| XM_005589356.2 | XP_005589413.1 | 2 - 3 | 566 | 73 | ||
| XM_015440936.1 | XP_015296422.1 | 2 - 3 | 569 | 73 | ||
| XM_015440937.1 | XP_015296423.1 | 2 - 3 | 296 | 73 | ||
| GenBank mRNA | AB169584.1 | - | 2 - 3 | 232 | 73 | |
| AB170222.1 | - | 2 - 3 | 224 | 73 | ||
| AB174290.1 | - | 2 - 3 | 228 | 73 | ||
More Information
Set Membership: |
|

DMAC2
Genomic Map