Assay Details
| Gene Symbol: CMTM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284611.1 | NP_001271540.1 | 1 - 2 | 159 | 68 | |
| GenBank mRNA | AB179205.1 | - | 1 - 2 | 159 | 68 | |
More Information
Set Membership: |
|

CMTM6
Genomic Map