Assay Details
| Gene Symbol: DERL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582652.2 | XP_005582709.1 | 6 - 7 | 661 | 107 | |
| XM_005582653.2 | XP_005582710.1 | 6 - 7 | 663 | 107 | ||
| XM_005582654.2 | XP_005582711.1 | 6 - 7 | 637 | 107 | ||
| XM_005582655.2 | XP_005582712.1 | 6 - 7 | 621 | 107 | ||
| XM_015437528.1 | XP_015293014.1 | 3 - 4 | 297 | 107 | ||
More Information
Set Membership: |
|

DERL2
Genomic Map