Assay Details
| Gene Symbol: SLC25A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582612.2 | XP_005582669.1 | 3 - 4 | 717 | 87 | |
| XM_005582614.2 | XP_005582671.1 | 3 - 4 | 489 | 87 | ||
| XM_015437510.1 | XP_015292996.1 | 2 - 3 | 703 | 87 | ||
More Information
Set Membership: |
|

SLC25A11
Genomic Map