Assay Details
| Gene Symbol: MRPS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557122.2 | XP_005557179.1 | 10 - 11 | 1057 | 104 | |
| XM_015451602.1 | XP_015307088.1 | 10 - 11 | 1018 | 104 | ||
More Information
Set Membership: |
|

MRPS27
Genomic Map