Assay Details
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283787.1 | NP_001270716.1 | 2 - 3 | 284 | 67 | |
XM_005576221.2 | XP_005576278.1 | 4 - 5 | 705 | 67 | ||
XM_005576222.2 | XP_005576279.1 | 3 - 4 | 644 | 67 | ||
XM_005576223.2 | XP_005576280.1 | 3 - 4 | 380 | 67 | ||
XM_015433430.1 | XP_015288916.1 | 2 - 3 | 1016 | 67 | ||
GenBank mRNA | AB062991.1 | - | 1 - 2 | 2502 | 67 | |
AB169838.1 | - | 2 - 3 | 283 | 67 | ||
AB170280.1 | - | 2 - 3 | 258 | 67 | ||
AB170332.1 | - | 2 - 3 | 284 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
