Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558311.2 | XP_005558368.1 | 7 - 8 | 699 | 62 | |
| GenBank mRNA | AB169483.1 | - | 7 - 8 | 667 | 62 | |
More Information
Set Membership: |
|

SPARC
Genomic Map