Assay Details
| Gene Symbol: GEMIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576073.2 | XP_005576130.1 | 2 - 3 | 307 | 101 | |
| XM_005576074.2 | XP_005576131.1 | 2 - 3 | 264 | 101 | ||
More Information
Set Membership: |
|

GEMIN6
Genomic Map