Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583311.2 | XP_005583368.2 | 2 - 3 | 177 | 107 | |
| XM_015438112.1 | XP_015293598.1 | 2 - 3 | 655 | 107 | ||
| XM_015438113.1 | XP_015293599.1 | 2 - 3 | 604 | 107 | ||
| XM_015438116.1 | XP_015293602.1 | 3 - 4 | 161 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015438112.1, XM_015438113.1, XM_015438116.1 transcripts.
Set Membership: |
|

NSRP1
Genomic Map