Assay Details
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005540348.1 | XP_005540405.1 | 6 - 7 | 1687 | 113 | |
XM_005540349.2 | XP_005540406.1 | 6 - 7 | 1572 | 113 | ||
XM_005540350.2 | XP_005540407.1 | 5 - 6 | 1597 | 113 | ||
XM_015447660.1 | XP_015303146.1 | 5 - 6 | 1435 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
