Assay Details
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542812.2 | XP_005542869.1 | 4 - 5 | 974 | 73 | |
XM_005542813.2 | XP_005542870.1 | 4 - 5 | 974 | 73 | ||
XM_005542814.2 | XP_005542871.1 | 5 - 6 | 698 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
