Assay Details
| Gene Symbol: MILR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584708.2 | XP_005584765.2 | 4 - 5 | 664 | 84 | |
| XM_015438713.1 | XP_015294199.1 | 4 - 5 | 666 | 84 | ||
More Information
Set Membership: |
|

MILR1
Genomic Map