Assay Details
| Gene Symbol: RSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283161.1 | NP_001270090.1 | 2 - 3 | 757 | 80 | |
| XM_015451683.1 | XP_015307169.1 | 1 - 2 | 607 | 80 | ||
| GenBank mRNA | AB169364.1 | - | 2 - 3 | 757 | 80 | |
More Information
Set Membership: |
|

RSRP1
Genomic Map