Assay Details
Gene Symbol: BEND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552679.2 | XP_005552736.1 | 6 - 7 | 1351 | 110 | |
XM_005552680.2 | XP_005552737.1 | 6 - 7 | 1029 | 110 | ||
XM_005552682.2 | XP_005552739.1 | 6 - 7 | 1026 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
