Assay Details
| Gene Symbol: GLIPR1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571574.2 | XP_005571631.1 | 2 - 3 | 517 | 80 | |
| XM_015431230.1 | XP_015286716.1 | 3 - 4 | 360 | 80 | ||
| XM_015431231.1 | XP_015286717.1 | 2 - 3 | 379 | 80 | ||
More Information
Set Membership: |
|

GLIPR1L2
Genomic Map