Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283770.1 | NP_001270699.1 | 2 - 3 | 714 | 61 | |
XM_015445341.1 | XP_015300827.1 | 2 - 3 | 1039 | 61 | ||
GenBank mRNA | AB063093.1 | - | 1265 | 61 | ||
AB169784.1 | - | 2 - 3 | 735 | 61 | ||
AB173479.1 | - | 2 - 3 | 714 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
