Assay Details
| Gene Symbol: TBRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580027.2 | XP_005580084.1 | 7 - 8 | 1192 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBRG1| Gene Symbol: TBRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580027.2 | XP_005580084.1 | 7 - 8 | 1192 | 92 | |
Set Membership: |
|