Assay Details
| Gene Symbol: METAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571898.2 | XP_005571955.1 | 5 - 6 | 1249 | 96 | |
| XM_005571899.2 | XP_005571956.1 | 5 - 6 | 682 | 96 | ||
| GenBank mRNA | AB171043.1 | - | 5 - 6 | 621 | 96 | |
More Information
Set Membership: |
|

METAP2
Genomic Map