Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572244.2 | XP_005572301.1 | 4 - 5 | 676 | 145 | |
| XM_005572245.1 | XP_005572302.1 | 4 - 5 | 598 | 145 | ||
| XM_005572247.2 | XP_005572304.1 | 4 - 5 | 613 | 145 | ||
| XM_005572248.2 | XP_005572305.1 | 3 - 4 | 511 | 145 | ||
| XM_015431528.1 | XP_015287014.1 | 3 - 4 | 439 | 145 | ||
| GenBank mRNA | AB578896.1 | - | 2 - 3 | 252 | 145 | |
| AB578897.1 | - | 2 - 3 | 252 | 145 | ||
| AB578898.1 | - | 2 - 3 | 252 | 145 | ||
More Information
Set Membership: |
|

VPS29
Genomic Map