Assay Details
| Gene Symbol: LOC102134040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591341.2 | XP_005591398.1 | 30 - 31 | 3696 | 61 | |
| XM_005591342.2 | XP_005591399.1 | 29 - 30 | 3576 | 61 | ||
| XM_015442577.1 | XP_015298063.1 | 30 - 31 | 3390 | 61 | ||
More Information
Set Membership: |
|

LOC102134040
Genomic Map