Assay Details
| Gene Symbol: RPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574156.1 | XP_005574213.1 | 1 - 2 | 215 | 65 | |
| XM_005574157.2 | XP_005574214.1 | 1 - 2 | 216 | 65 | ||
More Information
Set Membership: |
|

RPE
Genomic Map